Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639811.4 | XP_003639859.1 | 2 - 3 | 681 | 107 | |
| XM_005634065.3 | XP_005634122.1 | 2 - 3 | 726 | 107 | ||
| XM_005634066.3 | XP_005634123.1 | 2 - 3 | 319 | 107 | ||
More Information
Set Membership: |
|

POGLUT2
Genomic Map