Assay Details
| Gene Symbol: GADD45GIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533898.6 | XP_533898.2 | 1 - 2 | 385 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GADD45GIP1| Gene Symbol: GADD45GIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_533898.6 | XP_533898.2 | 1 - 2 | 385 | 54 | |
Set Membership: |
|