Assay Details
| Gene Symbol: GTF2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632979.3 | XP_005633036.1 | 10 - 11 | 1615 | 49 | |
| XM_005632980.3 | XP_005633037.1 | 10 - 11 | 1628 | 49 | ||
| XM_014121927.2 | XP_013977402.1 | 10 - 11 | 1606 | 49 | ||
| XM_022407135.1 | XP_022262843.1 | 10 - 11 | 1441 | 49 | ||
More Information
Set Membership: |
|

GTF2F1
Genomic Map