Assay Details
| Gene Symbol: DKK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284475.1 | NP_001271404.1 | 6 - 7 | 826 | 59 | |
| XM_014122426.2 | XP_013977901.1 | 6 - 7 | 655 | 59 | ||
| GenBank mRNA | AB733648.1 | - | 6 - 7 | 826 | 59 | |
More Information
Set Membership: |
|

DKK3
Genomic Map