Assay Details
| Gene Symbol: STX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108060.2 | XP_013963535.2 | 337 | 64 | ||
| XM_022410310.1 | XP_022266018.1 | 337 | 64 | |||
| XM_022410311.1 | XP_022266019.1 | 337 | 64 | |||
More Information
Set Membership: |
|

STX2
Genomic Map