Assay Details
Gene Symbol: LOC608697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106509.2 | XP_013961984.1 | 6 - 7 | 514 | 70 | |
XM_014106515.2 | XP_013961990.1 | 6 - 7 | 514 | 70 | ||
XM_022408603.1 | XP_022264311.1 | 6 - 7 | 514 | 70 | ||
XM_022408604.1 | XP_022264312.1 | 6 - 7 | 514 | 70 | ||
XM_022408605.1 | XP_022264313.1 | 5 - 6 | 469 | 70 | ||
XM_022408606.1 | XP_022264314.1 | 5 - 6 | 409 | 70 | ||
XM_022408607.1 | XP_022264315.1 | 5 - 6 | 437 | 70 | ||
XM_022408608.1 | XP_022264316.1 | 4 - 5 | 425 | 70 | ||
XM_022408609.1 | XP_022264317.1 | 4 - 5 | 364 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
