Assay Details
| Gene Symbol: MRPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639823.3 | XP_003639871.1 | 6 - 7 | 779 | 98 | |
| XM_022408628.1 | XP_022264336.1 | 6 - 7 | 780 | 98 | ||
| XM_022408629.1 | XP_022264337.1 | 6 - 7 | 780 | 98 | ||
| XM_022408630.1 | XP_022264338.1 | 6 - 7 | 782 | 98 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL3
Genomic Map