Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635459.3 | XP_005635516.1 | 1800 | 69 | ||
| XM_014107426.2 | XP_013962901.1 | 1794 | 69 | |||
| XM_022409585.1 | XP_022265293.1 | 1976 | 69 | |||
More Information
Set Membership: |
|

SPART
Genomic Map