Assay Details
| Gene Symbol: LOC477516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410295.1 | XP_022266003.1 | 9 - 10 | 1217 | 90 | |
| XM_022410296.1 | XP_022266004.1 | 8 - 9 | 981 | 90 | ||
| XM_022410297.1 | XP_022266005.1 | 4 - 5 | 908 | 90 | ||
| XM_022410298.1 | XP_022266006.1 | 4 - 5 | 340 | 90 | ||
| XM_534711.6 | XP_534711.3 | 5 - 6 | 3082 | 90 | ||
More Information
Set Membership: |
|

LOC477516
Genomic Map