Assay Details
| Gene Symbol: PTPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635173.3 | XP_005635230.1 | 7 - 8 | 1031 | 61 | |
| XM_005635174.3 | XP_005635231.1 | 7 - 8 | 1010 | 61 | ||
| XM_005635175.3 | XP_005635232.1 | 6 - 7 | 939 | 61 | ||
| XM_534455.6 | XP_534455.3 | 7 - 8 | 1030 | 61 | ||
More Information
Set Membership: |
|

PTPN1
Genomic Map