Assay Details
| Gene Symbol: UBE2F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635924.3 | XP_005635981.1 | 6 - 7 | 522 | 63 | |
| XM_534617.6 | XP_534617.3 | 6 - 7 | 516 | 63 | ||
More Information
Set Membership: |
|

UBE2F
Genomic Map