Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636216.2 | XP_005636273.1 | 537 | 122 | ||
| XM_005636217.2 | XP_005636274.1 | 498 | 122 | |||
More Information
Set Membership: |
|

FAM216A
Genomic Map