Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636279.3 | XP_005636336.1 | 7 - 8 | 1596 | 60 | |
| XM_534697.6 | XP_534697.3 | 7 - 8 | 1452 | 60 | ||
More Information
Set Membership: |
|

WSB2
Genomic Map