Assay Details
| Gene Symbol: LOC477556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410501.1 | XP_022266209.1 | 2 - 3 | 346 | 92 | |
| XM_534751.6 | XP_534751.3 | 3 - 4 | 435 | 92 | ||
| GenBank mRNA | KM233847.1 | - | 3 - 4 | 393 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022410501.1 transcripts.
Set Membership: |
|

LOC477556
Genomic Map