Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409168.1 | XP_022264876.1 | 3 - 4 | 360 | 58 | |
| XM_534405.6 | XP_534405.2 | 3 - 4 | 360 | 58 | ||
More Information
Set Membership: |
|

NFS1
Genomic Map