Assay Details
| Gene Symbol: RBIS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412267.1 | XP_022267975.1 | 3 - 4 | 255 | 97 | |
| XM_022412268.1 | XP_022267976.1 | 3 - 4 | 256 | 97 | ||
| XM_535116.7 | XP_535116.1 | 3 - 4 | 391 | 97 | ||
More Information
Set Membership: |
|

RBIS
Genomic Map