Assay Details
| Gene Symbol: TMBIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252399.1 | NP_001239328.1 | 3 - 4 | 234 | 60 | |
| XM_005636692.3 | XP_005636749.1 | 3 - 4 | 340 | 60 | ||
| XM_022410856.1 | XP_022266564.1 | 3 - 4 | 289 | 60 | ||
| GenBank mRNA | DN880529.1 | - | 3 - 4 | 234 | 60 | |
More Information
Set Membership: |
|

TMBIM6
Genomic Map