Assay Details
| Gene Symbol: RSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617104.3 | XP_005617161.1 | 595 | 68 | ||
| XM_022407439.1 | XP_022263147.1 | 447 | 68 | |||
| XM_022407441.1 | XP_022263149.1 | 647 | 68 | |||
| XM_022407443.1 | XP_022263151.1 | 647 | 68 | |||
| XM_535177.6 | XP_535177.1 | 646 | 68 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RSU1
Genomic Map