Assay Details
| Gene Symbol: CPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109125.2 | XP_013964600.1 | 8 - 9 | 1154 | 77 | |
| XM_534989.6 | XP_534989.4 | 7 - 8 | 1360 | 77 | ||
More Information
Set Membership: |
|

CPN1
Genomic Map