Assay Details
| Gene Symbol: ATP6V0D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535118.5 | XP_535118.2 | 6 - 7 | 1053 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V0D2| Gene Symbol: ATP6V0D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_535118.5 | XP_535118.2 | 6 - 7 | 1053 | 89 | |
Set Membership: |
|