Assay Details
| Gene Symbol: WASHC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108830.2 | XP_013964305.1 | 28 - 29 | 3212 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WASHC2C| Gene Symbol: WASHC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108830.2 | XP_013964305.1 | 28 - 29 | 3212 | 59 | |
Set Membership: |
|