Assay Details
| Gene Symbol: IMPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109269.2 | XP_013964744.1 | 4 - 5 | 425 | 84 | |
| XM_022412455.1 | XP_022268163.1 | 4 - 5 | 425 | 84 | ||
| XM_022412456.1 | XP_022268164.1 | 4 - 5 | 403 | 84 | ||
More Information
Set Membership: |
|

IMPA1
Genomic Map