Assay Details
| Gene Symbol: MYZAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638457.1 | XP_005638514.1 | 9 - 10 | 1023 | 80 | |
| XM_022412856.1 | XP_022268564.1 | 9 - 10 | 1023 | 80 | ||
More Information
Set Membership: |
|

MYZAP
Genomic Map