Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617174.3 | XP_005617231.1 | 1307 | 89 | ||
| XM_005617175.3 | XP_005617232.1 | 1282 | 89 | |||
| XM_022408084.1 | XP_022263792.1 | 1282 | 89 | |||
| XM_022408087.1 | XP_022263795.1 | 1144 | 89 | |||
| XM_022408090.1 | XP_022263798.1 | 1282 | 89 | |||
| XM_022408091.1 | XP_022263799.1 | 1282 | 89 | |||
More Information
Set Membership: |
|

TASOR2
Genomic Map