Assay Details
| Gene Symbol: LOC100856647 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640080.4 | XP_003640128.1 | 8 - 9 | 622 | 98 | |
| XM_022413179.1 | XP_022268887.1 | 8 - 9 | 1141 | 98 | ||
| XM_022413180.1 | XP_022268888.1 | 8 - 9 | 1059 | 98 | ||
| XM_022413181.1 | XP_022268889.1 | 8 - 9 | 622 | 98 | ||
| XM_022413182.1 | XP_022268890.1 | 8 - 9 | 622 | 98 | ||
| XM_022413183.1 | XP_022268891.1 | 7 - 8 | 858 | 98 | ||
| XM_022413184.1 | XP_022268892.1 | 7 - 8 | 776 | 98 | ||
| XM_022413185.1 | XP_022268893.1 | 7 - 8 | 611 | 98 | ||
| XM_022413186.1 | XP_022268894.1 | 7 - 8 | 1111 | 98 | ||
| XM_022413187.1 | XP_022268895.1 | 7 - 8 | 1029 | 98 | ||
| Gene Symbol: CRYZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413238.1 | XP_022268946.1 | 8 - 9 | 1141 | 98 | |
| XM_022413239.1 | XP_022268947.1 | 8 - 9 | 1059 | 98 | ||
| XM_022413240.1 | XP_022268948.1 | 8 - 9 | 622 | 98 | ||
| XM_022413241.1 | XP_022268949.1 | 8 - 9 | 622 | 98 | ||
| XM_022413242.1 | XP_022268950.1 | 7 - 8 | 859 | 98 | ||
| XM_022413243.1 | XP_022268951.1 | 7 - 8 | 777 | 98 | ||
| XM_022413244.1 | XP_022268952.1 | 7 - 8 | 611 | 98 | ||
| XM_022413245.1 | XP_022268953.1 | 7 - 8 | 1111 | 98 | ||
| XM_022413246.1 | XP_022268954.1 | 7 - 8 | 1029 | 98 | ||
| XM_535585.6 | XP_535585.1 | 8 - 9 | 622 | 98 | ||
More Information
Set Membership: |
|

LOC100856647
Genomic Map