Assay Details
| Gene Symbol: PAFAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617769.2 | XP_005617826.1 | 3 - 4 | 525 | 61 | |
| XM_005617770.2 | XP_005617827.1 | 3 - 4 | 491 | 61 | ||
| XM_005617771.2 | XP_005617828.1 | 3 - 4 | 525 | 61 | ||
| XM_005617772.2 | XP_005617829.1 | 3 - 4 | 528 | 61 | ||
| XM_005617773.3 | XP_005617830.1 | 4 - 5 | 899 | 61 | ||
| XM_005617774.3 | XP_005617831.1 | 3 - 4 | 808 | 61 | ||
| XM_005617777.3 | XP_005617834.1 | 3 - 4 | 484 | 61 | ||
| XM_014110437.2 | XP_013965912.1 | 4 - 5 | 986 | 61 | ||
| XM_014110439.2 | XP_013965914.1 | 4 - 5 | 968 | 61 | ||
| XM_535348.6 | XP_535348.2 | 4 - 5 | 575 | 61 | ||
More Information
Set Membership: |
|

PAFAH2
Genomic Map