Assay Details
| Gene Symbol: PPP1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113168.2 | NP_001106639.2 | 4 - 5 | 547 | 89 | |
| GenBank mRNA | BM539680.1 | - | 4 - 5 | 434 | 89 | |
More Information
Set Membership: |
|

PPP1R2
Genomic Map