Assay Details
| Gene Symbol: ITPRID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640411.3 | XP_005640468.2 | 12 - 13 | 2824 | 127 | |
| XM_014110785.2 | XP_013966260.1 | 14 - 15 | 3231 | 127 | ||
| XM_014110786.2 | XP_013966261.1 | 9 - 10 | 2633 | 127 | ||
| XM_014110787.2 | XP_013966262.1 | 12 - 13 | 2825 | 127 | ||
| XM_014110788.2 | XP_013966263.1 | 12 - 13 | 2827 | 127 | ||
| XM_022414834.1 | XP_022270542.1 | 12 - 13 | 2740 | 127 | ||
More Information
Set Membership: |
|

ITPRID2
Genomic Map