Assay Details
| Gene Symbol: MRPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110027.2 | XP_013965502.1 | 1 - 2 | 154 | 87 | |
| XM_535635.6 | XP_535635.1 | 1 - 2 | 154 | 87 | ||
More Information
Set Membership: |
|

MRPS18C
Genomic Map