Assay Details
| Gene Symbol: HPGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001200044.2 | NP_001186973.1 | 3 - 4 | 566 | 84 | |
| GenBank mRNA | AB564549.1 | - | 3 - 4 | 281 | 84 | |
More Information
Set Membership: |
|

HPGDS
Genomic Map