Assay Details
| Gene Symbol: SCARB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639077.3 | XP_005639134.1 | 7 - 8 | 1024 | 92 | |
| XM_022413552.1 | XP_022269260.1 | 7 - 8 | 1024 | 92 | ||
| XM_022413553.1 | XP_022269261.1 | 7 - 8 | 1071 | 92 | ||
| XM_022413554.1 | XP_022269262.1 | 6 - 7 | 817 | 92 | ||
More Information
Set Membership: |
|

SCARB2
Genomic Map