Assay Details
| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413546.1 | XP_022269254.1 | 5 - 6 | 649 | 82 | |
| XM_535703.6 | XP_535703.3 | 5 - 6 | 762 | 82 | ||
More Information
Set Membership: |
|

MYOZ2
Genomic Map