Assay Details
| Gene Symbol: SLC9B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109929.2 | XP_013965404.1 | 4 - 5 | 511 | 151 | |
| XM_014109930.2 | XP_013965405.1 | 5 - 6 | 579 | 151 | ||
| XM_014109931.2 | XP_013965406.1 | 6 - 7 | 725 | 151 | ||
| XM_014109932.2 | XP_013965407.1 | 5 - 6 | 635 | 151 | ||
| XM_014109933.2 | XP_013965408.1 | 4 - 5 | 441 | 151 | ||
| XM_014109935.2 | XP_013965410.1 | 3 - 4 | 371 | 151 | ||
| XM_022413401.1 | XP_022269109.1 | 3 - 4 | 286 | 151 | ||
| XM_022413402.1 | XP_022269110.1 | 4 - 5 | 517 | 151 | ||
More Information
Set Membership: |
|

SLC9B1
Genomic Map