Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639667.3 | XP_005639724.1 | 1233 | 90 | ||
| XM_022414192.1 | XP_022269900.1 | 1233 | 90 | |||
| XM_022414193.1 | XP_022269901.1 | 1277 | 90 | |||
| XM_022414194.1 | XP_022269902.1 | 1309 | 90 | |||
More Information
Set Membership: |
|

MARCHF6
Genomic Map