Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413636.1 | XP_022269344.1 | 2 - 3 | 151 | 123 | |
| XM_022413637.1 | XP_022269345.1 | 2 - 3 | 151 | 123 | ||
| XM_022413638.1 | XP_022269346.1 | 2 - 3 | 151 | 123 | ||
| XM_022413639.1 | XP_022269347.1 | 2 - 3 | 151 | 123 | ||
| XM_022413640.1 | XP_022269348.1 | 2 - 3 | 151 | 123 | ||
| XM_022413641.1 | XP_022269349.1 | 2 - 3 | 139 | 123 | ||
| XM_022413642.1 | XP_022269350.1 | 2 - 3 | 151 | 123 | ||
More Information
Set Membership: |
|

SEPTIN11
Genomic Map