Assay Details
Gene Symbol: CCNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639096.3 | XP_005639153.1 | 5 - 6 | 747 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
