Assay Details
| Gene Symbol: ABHD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110202.2 | XP_013965677.2 | 3 - 4 | 500 | 86 | |
| XM_535737.6 | XP_535737.5 | 3 - 4 | 490 | 86 | ||
More Information
Set Membership: |
|

ABHD10
Genomic Map