Assay Details
| Gene Symbol: ETFRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433562.4 | XP_003433610.1 | 3 - 4 | 332 | 113 | |
| XM_005637045.3 | XP_005637102.1 | 3 - 4 | 317 | 113 | ||
| XM_005637046.3 | XP_005637103.1 | 2 - 3 | 239 | 113 | ||
| XM_005637047.3 | XP_005637104.1 | 3 - 4 | 260 | 113 | ||
| XM_005637048.3 | XP_005637105.1 | 2 - 3 | 292 | 113 | ||
| XM_022411289.1 | XP_022266997.1 | 3 - 4 | 397 | 113 | ||
| XM_534862.6 | XP_534862.3 | 2 - 3 | 245 | 113 | ||
More Information
Set Membership: |
|

ETFRF1
Genomic Map