Assay Details
| Gene Symbol: STMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110454.2 | XP_013965929.1 | 3 - 4 | 523 | 75 | |
| XM_535349.6 | XP_535349.1 | 4 - 5 | 596 | 75 | ||
| XM_861033.5 | XP_866126.1 | 4 - 5 | 513 | 75 | ||
More Information
Set Membership: |
|

STMN1
Genomic Map