Assay Details
| Gene Symbol: ATP5IF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_860617.5 | XP_865710.1 | 2 - 3 | 255 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5IF1| Gene Symbol: ATP5IF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_860617.5 | XP_865710.1 | 2 - 3 | 255 | 61 | |
Set Membership: |
|