Assay Details
| Gene Symbol: ZNF567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115501.2 | XP_013970976.2 | 5 - 6 | 454 | 97 | |
| XM_014115506.2 | XP_013970981.1 | 5 - 6 | 722 | 97 | ||
| XM_022403715.1 | XP_022259423.1 | 6 - 7 | 556 | 97 | ||
| XM_022403718.1 | XP_022259426.1 | 6 - 7 | 586 | 97 | ||
| XM_022403719.1 | XP_022259427.1 | 6 - 7 | 552 | 97 | ||
| XM_022403722.1 | XP_022259430.1 | 5 - 6 | 391 | 97 | ||
| XM_022403730.1 | XP_022259438.1 | 5 - 6 | 479 | 97 | ||
More Information
Set Membership: |
|

ZNF567
Genomic Map