Assay Details
| Gene Symbol: ATF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638813.4 | XP_003638861.2 | 1 - 2 | 145 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003638813.4 transcripts.
Set Membership: |
|

ATF5
Genomic Map