Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633125.3 | XP_005633182.1 | 2 - 3 | 374 | 56 | |
| XM_005633127.2 | XP_005633184.1 | 2 - 3 | 433 | 56 | ||
| XM_022407269.1 | XP_022262977.1 | 2 - 3 | 374 | 56 | ||
| XM_022407270.1 | XP_022262978.1 | 2 - 3 | 372 | 56 | ||
| XM_022407271.1 | XP_022262979.1 | 2 - 3 | 682 | 56 | ||
| XM_022407272.1 | XP_022262980.1 | 2 - 3 | 580 | 56 | ||
| XM_022407273.1 | XP_022262981.1 | 2 - 3 | 433 | 56 | ||
| XM_022407274.1 | XP_022262982.1 | 2 - 3 | 490 | 56 | ||
| XM_022407275.1 | XP_022262983.1 | 2 - 3 | 374 | 56 | ||
More Information
Set Membership: |
|

MBD3
Genomic Map