Assay Details
| Gene Symbol: ZNF428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425283.1 | XP_022280991.1 | 2 - 3 | 589 | 88 | |
| XM_022425288.1 | XP_022280996.1 | 2 - 3 | 404 | 88 | ||
| XM_847576.5 | XP_852669.1 | 2 - 3 | 557 | 88 | ||
More Information
Set Membership: |
|

ZNF428
Genomic Map