Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629294.3 | XP_005629351.1 | 1 - 2 | 161 | 127 | |
| XM_005629295.3 | XP_005629352.1 | 1 - 2 | 161 | 127 | ||
| XM_022403429.1 | XP_022259137.1 | 1 - 2 | 164 | 127 | ||
| XM_532668.6 | XP_532668.1 | 1 - 2 | 161 | 127 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WASHC3
Genomic Map