Assay Details
| Gene Symbol: MIDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633148.2 | XP_005633205.1 | 1 - 2 | 468 | 80 | |
| XM_005633149.2 | XP_005633206.1 | 1 - 2 | 468 | 80 | ||
| XM_014122131.2 | XP_013977606.1 | 1 - 2 | 468 | 80 | ||
| XM_014122132.2 | XP_013977607.1 | 1 - 2 | 468 | 80 | ||
| XM_014122133.2 | XP_013977608.1 | 1 - 2 | 468 | 80 | ||
| XM_863512.4 | XP_868605.1 | 1 - 2 | 468 | 80 | ||
More Information
Set Membership: |
|

MIDN
Genomic Map