Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619287.3 | XP_005619344.1 | 5 - 6 | 812 | 64 | |
| XM_005619288.3 | XP_005619345.1 | 4 - 5 | 657 | 64 | ||
| XM_005619289.3 | XP_005619346.1 | 3 - 4 | 869 | 64 | ||
| XM_005619290.3 | XP_005619347.1 | 3 - 4 | 522 | 64 | ||
| XM_005619291.3 | XP_005619348.1 | 4 - 5 | 677 | 64 | ||
| XM_005619292.3 | XP_005619349.1 | 4 - 5 | 653 | 64 | ||
| XM_005619293.3 | XP_005619350.1 | 4 - 5 | 630 | 64 | ||
| XM_005619294.3 | XP_005619351.1 | 3 - 4 | 531 | 64 | ||
| XM_005619295.2 | XP_005619352.1 | 4 - 5 | 680 | 64 | ||
More Information
Set Membership: |
|

MYOZ3
Genomic Map