Assay Details
| Gene Symbol: OAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637828.2 | XP_005637885.1 | 9 - 10 | 1232 | 97 | |
| XM_014109077.2 | XP_013964552.1 | 10 - 11 | 1449 | 97 | ||
| XM_022411766.1 | XP_022267474.1 | 9 - 10 | 1173 | 97 | ||
| XM_022411767.1 | XP_022267475.1 | 9 - 10 | 1221 | 97 | ||
| XM_022411769.1 | XP_022267477.1 | 8 - 9 | 1004 | 97 | ||
| XM_860971.4 | XP_866064.1 | 10 - 11 | 1401 | 97 | ||
More Information
Set Membership: |
|

OAT
Genomic Map