Assay Details
| Gene Symbol: MSMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313785.1 | NP_001300714.1 | 4 - 5 | 640 | 96 | |
| XM_005629392.3 | XP_005629449.1 | 4 - 5 | 1924 | 96 | ||
More Information
Set Membership: |
|

MSMO1
Genomic Map