Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122027.1 | XP_013977502.1 | 3 - 4 | 810 | 73 | |
| XM_847807.5 | XP_852900.1 | 2 - 3 | 887 | 73 | ||
More Information
Set Membership: |
|

DNAJB1
Genomic Map